Search
Search

pqn-36| Gene Symbol: pqn-36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001364051.5 | NP_001350994.1 | 9 | 2109 | 92 | |
| NM_001364053.4 | NP_001350996.1 | 10 - 11 | 2124 | 92 | ||
| NM_078144.4 | NP_510545.1 | 8 - 9 | 2912 | 92 | ||
| NM_078146.5 | NP_510547.1 | 8 - 9 | 2655 | 92 | ||
Set Membership: |
|