Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: catp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129056.5 | NP_001122528.1 | 15 - 16 | 3252 | 82 | |
GenBank mRNA | AM419898.1 | - | 15 - 16 | 3241 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |