Search
Search

unc-64| Gene Symbol: unc-64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027443.5 | NP_001022614.1 | 3 - 4 | 317 | 108 | |
| NM_001379980.1 | NP_001366988.1 | 3 - 4 | 319 | 108 | ||
| GenBank mRNA | AB008842.1 | - | 3 - 4 | 327 | 108 | |
| AB008843.1 | - | 3 - 4 | 327 | 108 | ||
| AB008844.1 | - | 3 - 4 | 327 | 108 | ||
| AF047885.1 | - | 3 - 4 | 329 | 108 | ||
Set Membership: |
|