Search
Search

| Gene Symbol: egl-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028052.5 | NP_001023223.1 | 6 - 7 | 1059 | 86 | |
| NM_001307521.4 | NP_001294450.1 | 6 - 7 | 953 | 86 | ||
| NM_001380068.1 | NP_001368271.1 | 1 - 2 | 65 | 86 | ||
| NM_067740.6 | NP_500141.1 | 6 - 7 | 953 | 86 | ||
| NM_067741.5 | NP_500142.1 | 6 - 7 | 824 | 86 | ||
| NM_171279.6 | NP_741329.1 | 6 - 7 | 860 | 86 | ||
| NM_171280.4 | NP_741330.2 | 2 - 3 | 389 | 86 | ||
| Gene Symbol: ccm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380067.2 | NP_001367894.1 | 15 - 16 | 1851 | 86 | |
Set Membership: |
|