Search
Search

ztf-3| Gene Symbol: ztf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306291.3 | NP_001293220.1 | 1 - 2 | 389 | 90 | |
| NM_001373713.1 | NP_001360315.1 | 4 - 5 | 950 | 90 | ||
| NM_001381400.1 | NP_001368396.1 | 4 - 5 | 800 | 90 | ||
Set Membership: |
|