Search
Search

mab-20| Gene Symbol: mab-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306514.3 | NP_001293443.1 | 4 - 5 | 720 | 60 | |
| NM_001306515.3 | NP_001293444.1 | 3 - 4 | 549 | 60 | ||
| NM_058474.5 | NP_490875.1 | 5 - 6 | 1282 | 60 | ||
| NR_165326.1 | 4 - 5 | 1967 | 60 | |||
Set Membership: |
|