Search
Search

scm-1| Gene Symbol: scm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306425.3 | NP_001293354.1 | 1 - 2 | 110 | 72 | |
| NM_058510.7 | NP_490911.1 | 5 - 6 | 1432 | 72 | ||
| GenBank mRNA | AF225921.1 | - | 3 - 4 | 342 | 72 | |
Set Membership: |
|