Search
Search

unc-105| Gene Symbol: unc-105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129123.3 | NP_001122595.1 | 9 - 10 | 2367 | 108 | |
| NM_001377828.2 | NP_001364812.1 | 10 - 11 | 2355 | 108 | ||
| NM_001381517.1 | NP_001368112.1 | 10 - 11 | 2349 | 108 | ||
| NM_001381518.2 | NP_001367274.1 | 10 - 11 | 2349 | 108 | ||
| NM_063301.6 | NP_495702.1 | 9 - 10 | 2167 | 108 | ||
Set Membership: |
|