Search
Search

unc-93| Gene Symbol: unc-93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026047.5 | NP_001021218.1 | 11 - 12 | 1426 | 126 | |
| NM_001381807.1 | NP_001366876.1 | 11 - 12 | 1414 | 126 | ||
Set Membership: |
|