Search
Search

ndg-4| Gene Symbol: ndg-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027447.6 | NP_001022618.1 | 11 - 12 | 1917 | 58 | |
| NM_001027448.4 | NP_001022619.1 | 11 - 12 | 1911 | 58 | ||
| GenBank mRNA | AF173373.1 | - | 11 - 12 | 1915 | 58 | |
Set Membership: |
|