Search
Search

nep-16| Gene Symbol: nep-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028032.6 | NP_001023203.2 | 9 - 10 | 1284 | 151 | |
| NM_001028034.5 | NP_001023205.2 | 7 - 8 | 948 | 151 | ||
| NM_001383127.1 | 8 - 9 | 1080 | 151 | |||
Set Membership: |
|