Search
Search

smgl-2| Gene Symbol: smgl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307445.3 | NP_001294374.1 | 5 - 6 | 860 | 137 | |
| NM_001380158.1 | NP_001368241.1 | 1 - 2 | 125 | 137 | ||
Set Membership: |
|