Search
Search

pqn-22| Gene Symbol: pqn-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306867.3 | NP_001293796.1 | 3 - 4 | 788 | 96 | |
| NM_001306868.2 | NP_001293797.1 | 3 - 4 | 788 | 96 | ||
| NM_001306869.4 | NP_001293798.1 | 3 - 4 | 788 | 96 | ||
| NM_001306870.4 | NP_001293799.1 | 3 - 4 | 788 | 96 | ||
| NM_001306871.3 | NP_001293800.1 | 3 - 4 | 788 | 96 | ||
| NM_001306872.3 | NP_001293801.1 | 3 - 4 | 788 | 96 | ||
| NM_001306873.3 | NP_001293802.1 | 3 - 4 | 788 | 96 | ||
| NM_001306874.3 | NP_001293803.1 | 4 - 5 | 1043 | 96 | ||
| NM_001306875.3 | NP_001293804.1 | 4 - 5 | 1043 | 96 | ||
| NM_001306876.3 | NP_001293805.1 | 4 - 5 | 1043 | 96 | ||
| NM_001306877.4 | NP_001293806.1 | 4 - 5 | 1043 | 96 | ||
| NM_001306878.3 | NP_001293807.1 | 4 - 5 | 1043 | 96 | ||
| NM_001306879.3 | NP_001293808.1 | 4 - 5 | 1043 | 96 | ||
| NM_001306880.3 | NP_001293809.1 | 4 - 5 | 1043 | 96 | ||
| NM_068505.4 | NP_500906.2 | 3 - 4 | 788 | 96 | ||
| NM_068506.3 | NP_500907.2 | 4 - 5 | 1043 | 96 | ||
Set Membership: |
|