Search
Search

kin-4| Gene Symbol: kin-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268622.2 | NP_001255551.1 | 11 - 12 | 2472 | 67 | |
| NM_001268624.3 | NP_001255553.1 | 11 - 12 | 2463 | 67 | ||
| NM_001268625.3 | NP_001255554.1 | 10 - 11 | 1926 | 67 | ||
| NM_001268626.3 | NP_001255555.1 | 9 - 10 | 1914 | 67 | ||
| NM_001268627.2 | NP_001255556.1 | 11 - 12 | 2463 | 67 | ||
| NM_001268629.3 | NP_001255558.1 | 6 - 7 | 1125 | 67 | ||
| NM_001307048.3 | NP_001293977.1 | 8 - 9 | 1683 | 67 | ||
| NM_001380461.1 | NP_001367231.1 | 10 - 11 | 2367 | 67 | ||
| NM_001380462.2 | NP_001366913.1 | 10 - 11 | 2406 | 67 | ||
| GenBank mRNA | AM086627.1 | - | 8 - 9 | 1674 | 67 | |
Set Membership: |
|