Search
Search

tag-273| Gene Symbol: tag-273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028346.4 | NP_001023517.2 | 1 - 2 | 131 | 64 | |
| NM_001268884.3 | NP_001255813.1 | 1 - 2 | 104 | 64 | ||
| NM_001307072.3 | NP_001294001.1 | 1 - 2 | 104 | 64 | ||
Set Membership: |
|