Search
Search

gcy-25| Gene Symbol: gcy-25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307187.2 | NP_001294116.1 | 2 - 3 | 354 | 101 | |
| NM_001307188.3 | NP_001294117.1 | 2 - 3 | 327 | 101 | ||
Set Membership: |
|