Search
Search

daf-11| Gene Symbol: daf-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313082.3 | NP_001300011.1 | 11 - 12 | 2135 | 149 | |
| NM_073559.8 | NP_505960.3 | 16 - 17 | 3157 | 149 | ||
| NR_132532.1 | 15 - 16 | 2794 | 149 | |||
Set Membership: |
|