Search
Search

gpcp-1| Gene Symbol: gpcp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029613.4 | NP_001024784.2 | 2 - 3 | 279 | 119 | |
| NM_001270069.5 | NP_001256998.1 | 2 - 3 | 266 | 119 | ||
| NM_001380928.2 | NP_001367507.1 | 2 - 3 | 266 | 119 | ||
Set Membership: |
|