Search
Search

dnj-14| Gene Symbol: dnj-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270085.3 | NP_001257014.1 | 1 - 2 | 181 | 135 | |
| NM_001270086.3 | NP_001257015.1 | 1 - 2 | 175 | 135 | ||
Set Membership: |
|