Search
Search

him-4| Gene Symbol: him-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029411.3 | NP_001024582.1 | 54 - 55 | 13555 | 70 | |
| NM_001373450.2 | NP_001360633.1 | 53 - 54 | 13200 | 70 | ||
| NM_001373451.2 | NP_001360634.1 | 53 - 54 | 13200 | 70 | ||
| NM_077234.7 | NP_509635.2 | 54 - 55 | 13494 | 70 | ||
| GenBank mRNA | AF074901.1 | - | 54 - 55 | 13494 | 70 | |
Set Membership: |
|