Search
Search
| Gene Symbol: PRRC2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627136.3 | XP_005627193.1 | 6 - 7 | 821 | 84 | |
| XM_014118308.2 | XP_013973783.1 | 6 - 7 | 821 | 84 | ||
| XM_022425833.1 | XP_022281541.1 | 6 - 7 | 822 | 84 | ||
| XM_022425834.1 | XP_022281542.1 | 6 - 7 | 822 | 84 | ||
Set Membership: |
|