Search
Search

PNLIPRP1| Gene Symbol: PNLIPRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003319.2 | NP_001003319.2 | 10 - 11 | 1090 | 77 | |
| GenBank mRNA | M35302.1 | - | 10 - 11 | 1096 | 77 | |
Set Membership: |
|