Search
Search
| Gene Symbol: PFKM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003199.1 | NP_001003199.1 | 4 - 5 | 452 | 85 | |
| GenBank mRNA | JQ250035.1 | - | 6 - 7 | 643 | 85 | |
| U25183.1 | - | 4 - 5 | 452 | 85 | ||
Set Membership: |
|