Search
Search
| Gene Symbol: ATP11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634110.3 | XP_005634167.1 | 17 - 18 | 2016 | 80 | |
| XM_005634111.3 | XP_005634168.1 | 17 - 18 | 2015 | 80 | ||
| XM_005634112.3 | XP_005634169.1 | 17 - 18 | 2015 | 80 | ||
| XM_005634113.3 | XP_005634170.1 | 16 - 17 | 1820 | 80 | ||
| XM_014106452.2 | XP_013961927.1 | 17 - 18 | 2016 | 80 | ||
| XM_014106453.2 | XP_013961928.1 | 17 - 18 | 2017 | 80 | ||
| XM_014106454.2 | XP_013961929.1 | 17 - 18 | 2016 | 80 | ||
| XM_022408165.1 | XP_022263873.1 | 16 - 17 | 1888 | 80 | ||
Set Membership: |
|