Search
Search

LOC476416| Gene Symbol: LOC476416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014118931.2 | XP_013974406.1 | 3 - 4 | 735 | 76 | |
| XM_014118940.2 | XP_013974415.1 | 3 - 4 | 733 | 76 | ||
| XM_022423976.1 | XP_022279684.1 | 3 - 4 | 736 | 76 | ||
| XM_022423979.1 | XP_022279687.1 | 5 - 6 | 1206 | 76 | ||
| XM_022423981.1 | XP_022279689.1 | 4 - 5 | 897 | 76 | ||
| XM_022423986.1 | XP_022279694.1 | 4 - 5 | 913 | 76 | ||
| XM_022423988.1 | XP_022279696.1 | 4 - 5 | 840 | 76 | ||
| XM_022423994.1 | XP_022279702.1 | 3 - 4 | 1078 | 76 | ||
| XM_022423997.1 | XP_022279705.1 | 2 - 3 | 827 | 76 | ||
| XM_022424006.1 | XP_022279714.1 | 3 - 4 | 733 | 76 | ||
Set Membership: |
|