Search
Search

MOV10L1| Gene Symbol: MOV10L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625719.2 | XP_005625776.1 | 2 - 3 | 333 | 77 | |
| XM_005625721.3 | XP_005625778.1 | 2 - 3 | 334 | 77 | ||
| XM_014117264.2 | XP_013972739.1 | 2 - 3 | 333 | 77 | ||
| XM_014117266.2 | XP_013972741.1 | 2 - 3 | 334 | 77 | ||
| XM_022424265.1 | XP_022279973.1 | 2 - 3 | 334 | 77 | ||
| XM_022424266.1 | XP_022279974.1 | 2 - 3 | 334 | 77 | ||
| XM_022424267.1 | XP_022279975.1 | 2 - 3 | 325 | 77 | ||
Set Membership: |
|