Search
Search
| Gene Symbol: DGUOK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404809.1 | XP_022260517.1 | 4 - 5 | 621 | 120 | |
| XM_022404810.1 | XP_022260518.1 | 5 - 6 | 666 | 120 | ||
| XM_533001.6 | XP_533001.3 | 6 - 7 | 880 | 120 | ||
Set Membership: |
|