Search
Search
| Gene Symbol: ME3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633384.3 | XP_005633441.1 | 11 - 12 | 1515 | 79 | |
| XM_022407544.1 | XP_022263252.1 | 10 - 11 | 1354 | 79 | ||
| XM_022407545.1 | XP_022263253.1 | 10 - 11 | 1083 | 79 | ||
| XM_022407546.1 | XP_022263254.1 | 10 - 11 | 1114 | 79 | ||
Set Membership: |
|