Search
Search
| Gene Symbol: RAB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313859.1 | NP_001300788.1 | 1 - 2 | 145 | 117 | |
| XM_005622800.3 | XP_005622857.1 | 1 - 2 | 64 | 117 | ||
| XM_014114983.2 | XP_013970458.1 | 1 - 2 | 244 | 117 | ||
| XM_022420771.1 | XP_022276479.1 | 1 - 2 | 243 | 117 | ||
Set Membership: |
|