Search
Search
| Gene Symbol: KLHL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628620.3 | XP_005628677.1 | 8 - 9 | 989 | 118 | |
| XM_005628621.3 | XP_005628678.1 | 8 - 9 | 1306 | 118 | ||
| XM_022427469.1 | XP_022283177.1 | 9 - 10 | 1589 | 118 | ||
| XM_022427471.1 | XP_022283179.1 | 8 - 9 | 1248 | 118 | ||
| XM_022427472.1 | XP_022283180.1 | 7 - 8 | 840 | 118 | ||
| XM_532490.6 | XP_532490.4 | 7 - 8 | 1176 | 118 | ||
Set Membership: |
|