Search
Search
| Gene Symbol: PUF60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628044.2 | XP_005628101.1 | 6 - 7 | 629 | 152 | |
| XM_005628045.2 | XP_005628102.1 | 5 - 6 | 457 | 152 | ||
| XM_005628046.2 | XP_005628103.1 | 5 - 6 | 536 | 152 | ||
| XM_005628047.2 | XP_005628104.1 | 6 - 7 | 1070 | 152 | ||
| XM_005628049.3 | XP_005628106.1 | 4 - 5 | 370 | 152 | ||
| XM_843617.5 | XP_848710.2 | 6 - 7 | 633 | 152 | ||
| XM_851947.4 | XP_857040.1 | 5 - 6 | 582 | 152 | ||
Set Membership: |
|