Search
Search
| Gene Symbol: SEMA4F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630511.3 | XP_005630568.2 | 4 - 5 | 902 | 90 | |
| XM_005630513.3 | XP_005630570.2 | 4 - 5 | 902 | 90 | ||
| XM_022404779.1 | XP_022260487.1 | 4 - 5 | 902 | 90 | ||
| XM_022404780.1 | XP_022260488.1 | 3 - 4 | 403 | 90 | ||
| XM_540214.6 | XP_540214.4 | 4 - 5 | 902 | 90 | ||
Set Membership: |
|