Search
Search
| Gene Symbol: GGCX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630488.3 | XP_005630545.1 | 9 - 10 | 1379 | 122 | |
| XM_014120376.2 | XP_013975851.1 | 7 - 8 | 1133 | 122 | ||
| XM_532979.6 | XP_532979.3 | 9 - 10 | 1374 | 122 | ||
Set Membership: |
|