Search
Search

BLOC1S2| Gene Symbol: BLOC1S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637614.3 | XP_005637671.1 | 4 - 5 | 486 | 130 | |
| XM_005637615.3 | XP_005637672.1 | 4 - 5 | 486 | 130 | ||
| XM_846354.5 | XP_851447.1 | 4 - 5 | 457 | 130 | ||
Set Membership: |
|