Search
Search
| Gene Symbol: DPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287158.1 | NP_001274087.1 | 3 | 527 | 185 | |
| GenBank mRNA | DN379133.1 | - | 3 | 527 | 185 | |
| DN432131.1 | - | 2 | 165 | 185 | ||
Set Membership: |
|