Search
Search
| Gene Symbol: LMO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420532.1 | XP_022276240.1 | 3 - 4 | 401 | 134 | |
| XM_537086.4 | XP_537086.1 | 3 - 4 | 939 | 134 | ||
| XM_862220.5 | XP_867313.1 | 3 - 4 | 495 | 134 | ||
Set Membership: |
|