Search
Search
| Gene Symbol: AKAP8L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022406947.1 | XP_022262655.1 | 12 - 13 | 1404 | 103 | |
| XM_022406948.1 | XP_022262656.1 | 12 - 13 | 1485 | 103 | ||
| XM_847928.5 | XP_853021.4 | 12 - 13 | 1668 | 103 | ||
Set Membership: |
|