Search
Search

TMEM161B| Gene Symbol: TMEM161B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618138.3 | XP_005618195.1 | 1364 | 178 | ||
| XM_005618139.3 | XP_005618196.1 | 1385 | 178 | |||
| XM_005618140.3 | XP_005618197.1 | 1373 | 178 | |||
| XM_005618141.3 | XP_005618198.1 | 1265 | 178 | |||
| XM_014112233.2 | XP_013967708.1 | 1306 | 178 | |||
| XM_022416738.1 | XP_022272446.1 | 1187 | 178 | |||
| XM_022416739.1 | XP_022272447.1 | 1394 | 178 | |||
| XM_022416740.1 | XP_022272448.1 | 1202 | 178 | |||
| XM_022416741.1 | XP_022272449.1 | 1311 | 178 | |||
| XM_022416742.1 | XP_022272450.1 | 1191 | 178 | |||
| XM_022416743.1 | XP_022272451.1 | 1445 | 178 | |||
| XM_022416744.1 | XP_022272452.1 | 1454 | 178 | |||
| XM_022416745.1 | XP_022272453.1 | 1485 | 178 | |||
| XM_022416746.1 | XP_022272454.1 | 840 | 178 | |||
| XM_546033.6 | XP_546033.4 | 1311 | 178 | |||
Set Membership: |
|