Search
Search
| Gene Symbol: SRSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638895.3 | XP_003638943.1 | 6 | 938 | 135 | |
| XM_003638896.4 | XP_003638944.1 | 6 | 1173 | 135 | ||
| XM_005617807.2 | XP_005617864.1 | 6 | 937 | 135 | ||
| XM_005617808.3 | XP_005617865.1 | 6 | 1171 | 135 | ||
| XM_014110637.2 | XP_013966112.1 | 7 | 1042 | 135 | ||
| XM_861323.4 | XP_866416.2 | 5 | 885 | 135 | ||
Set Membership: |
|