Search
Search

Sep15| Gene Symbol: Sep15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256866.1 | NP_001243795.1 | 2 - 3 | 328 | 98 | |
| GenBank mRNA | JI893016.1 | - | 2 - 3 | 328 | 98 | |
| JP051733.1 | - | 2 - 3 | 328 | 98 | ||