Search
Search

Mrc1| Gene Symbol: Mrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503375.1 | XP_003503423.1 | 15 - 16 | 2337 | 64 | |
| XM_007643986.1 | XP_007642176.1 | 15 - 16 | 2337 | 64 | ||
Set Membership: |
|