Search
Search

Slc5a10| Gene Symbol: Slc5a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496337.2 | XP_003496385.1 | 2 - 3 | 186 | 61 | |
| XM_007649266.1 | XP_007647456.1 | 2 - 3 | 186 | 61 | ||
| XM_007649269.1 | XP_007647459.1 | 2 - 3 | 186 | 61 | ||
Set Membership: |
|