Search
Search

Hmgxb4| Gene Symbol: Hmgxb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511959.2 | XP_003512007.1 | 6 - 7 | 2134 | 137 | |
| XM_007652470.1 | XP_007650660.1 | 7 - 8 | 1456 | 137 | ||
| XM_007652471.1 | XP_007650661.1 | 7 - 8 | 1873 | 137 | ||
| XM_007652472.1 | XP_007650662.1 | 6 - 7 | 1447 | 137 | ||
Set Membership: |
|