Search
Search

Sfpq| Gene Symbol: Sfpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641596.1 | XP_007639786.1 | 1 - 2 | 287 | 63 | |
| GenBank mRNA | JI886598.1 | - | 1 - 2 | 287 | 63 | |
| JP043418.1 | - | 1 - 2 | 287 | 63 | ||
Set Membership: |
|