Search
Search

Sel1l3| Gene Symbol: Sel1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512097.2 | XP_003512145.2 | 16 - 17 | 2448 | 90 | |
| GenBank mRNA | JI874091.1 | - | 4 - 5 | 486 | 90 | |
| JP062754.1 | - | 4 - 5 | 486 | 90 | ||