Search
Search

Mllt4| Gene Symbol: Mllt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644575.1 | XP_007642765.1 | 10 - 11 | 1497 | 59 | |
| XM_007644576.1 | XP_007642766.1 | 10 - 11 | 1500 | 59 | ||
| XM_007644577.1 | XP_007642767.1 | 5 - 6 | 718 | 59 | ||
Set Membership: |
|