Search
Search

Agbl3| Gene Symbol: Agbl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508900.2 | XP_003508948.2 | 4 - 5 | 514 | 76 | |
| XM_007649514.1 | XP_007647704.1 | 4 - 5 | 514 | 76 | ||
| XM_007649515.1 | XP_007647705.1 | 4 - 5 | 424 | 76 | ||
| XM_007649516.1 | XP_007647706.1 | 4 - 5 | 514 | 76 | ||
| XM_007649517.1 | XP_007647707.1 | 2 - 3 | 311 | 76 | ||
| XM_007649518.1 | XP_007647708.1 | 3 - 4 | 297 | 76 | ||
| XM_007649519.1 | XP_007647709.1 | 4 - 5 | 514 | 76 | ||
| XM_007649520.1 | XP_007647710.1 | 2 - 3 | 208 | 76 | ||
| XM_007649521.1 | XP_007647711.1 | 4 - 5 | 514 | 76 | ||
| GenBank mRNA | JI882950.1 | - | 2 - 3 | 235 | 76 | |
| JP051235.1 | - | 2 - 3 | 235 | 76 | ||
Set Membership: |
|