Search
Search

Gabarapl1| Gene Symbol: Gabarapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509252.2 | XP_003509300.1 | 1 - 2 | 361 | 105 | |
| GenBank mRNA | JI866515.1 | - | 1 - 2 | 345 | 105 | |
| JP064134.1 | - | 1 - 2 | 345 | 105 | ||
Set Membership: |
|