Search
Search

Gtpbp8| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497855.2 | XP_003497903.1 | 2 - 3 | 414 | 102 | |
| XM_007618707.1 | XP_007616897.1 | 2 - 3 | 414 | 102 | ||
| XM_007618715.1 | XP_007616905.1 | 2 - 3 | 414 | 102 | ||
Set Membership: |
|