Search
Search

Chtf18| Gene Symbol: Chtf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642156.1 | XP_007640346.1 | 13 - 14 | 1780 | 93 | |
| XM_007642157.1 | XP_007640347.1 | 13 - 14 | 1778 | 93 | ||
| GenBank mRNA | JI888968.1 | - | 3 - 4 | 213 | 93 | |
| JP061985.1 | - | 3 - 4 | 213 | 93 | ||
Set Membership: |
|